accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONOCl

CAS RN 14432-16-7

IUPAC Name: 2-chloro-4-nitropyridin-1-ium-1-olate
Synonyms: 2-chloro-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol): 174.54
Molecular Formula: C5H3ClN2O3
InChi Key: YSTCMHHKDOVZDA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=[N+]([O-])C(Cl)=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results