CAS RN 14435-88-2
IUPAC Name:
2,6-bis(4-methylphenyl)pyridine
Synonyms:
2,6-bis(4-methylphenyl)pyridine
Molecular Weight (g/mol):
259.35
Molecular Formula:
C19H17N
InChi Key:
BLRYHCTZTSRBNB-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=CC=CC(=N1)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
Promotions Available
2,6-Bis(p-tolyl)pyridine, 97%
2,6-Bis(p-tolyl)pyridine, 97%
CAS: 14435-88-2 Molecular Formula: C19H17N Molecular Weight (g/mol): 259.352 MDL Number: MFCD00006282 InChI Key: BLRYHCTZTSRBNB-UHFFFAOYSA-N PubChem CID: 84435 IUPAC Name: 2,6-bis(4-methylphenyl)pyridine SMILES: CC1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=C(C=C3)C