accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOOH

CAS RN 14440-94-9

IUPAC Name: 3-propoxypyridine-2-carboxylic acid
Synonyms: 3-propoxypyridine-2-carboxylic acid
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: XJFABQUUYAJFFJ-UHFFFAOYSA-N
SMILES: CCCOC1=C(N=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results