accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14442-12-7

IUPAC Name: 3-phenyl-1,2-oxazole-5-carboxylate
Synonyms: 3-phenyl-1,2-oxazole-5-carboxylate
Molecular Weight (g/mol): 188.16
Molecular Formula: C10H6NO3
InChi Key: YGTFDJRCCBKLDM-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=NO1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)