CAS RN 14447-15-5
CAS RN 14447-15-5
IUPAC Name:
propyl 2-cyanoacetate
Synonyms:
propyl 2-cyanoacetate
Molecular Weight (g/mol):
127.14
Molecular Formula:
C6H9NO2
InChi Key:
NLFIMXLLXGTDME-UHFFFAOYSA-N
SMILES:
CCCOC(=O)CC#N
1
–
1
of
1
results
1
Propyl Cyanoacetate 99.0+%, TCI America™
CAS: 14447-15-5 Molecular Formula: C6H9NO2 Molecular Weight (g/mol): 127.14 MDL Number: MFCD00015784 InChI Key: NLFIMXLLXGTDME-UHFFFAOYSA-N Synonym: Cyanoacetic Acid Propyl Ester PubChem CID: 84444 IUPAC Name: propyl 2-cyanoacetate SMILES: CCCOC(=O)CC#N