accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14452-34-7

IUPAC Name: 2,4-dimethyl-6-nitrophenol
Synonyms: 2,4-dimethyl-6-nitrophenol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: KJRCHILWKQLEBC-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(O)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)