accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 144584-66-7

FFOBrO

CAS RN 144584-66-7

IUPAC Name: 4-bromo-2,2-difluoro-2H-1,3-benzodioxole
Synonyms: 4-bromo-2,2-difluoro-1,3-benzodioxole
Molecular Weight (g/mol): 237
Molecular Formula: C7H3BrF2O2
InChi Key: LSZYHXNOLVSZHH-UHFFFAOYSA-N
SMILES: FC1(F)OC2=CC=CC(Br)=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results