accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1446-61-3

IUPAC Name: [(1R,4aS,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl]methanaminium
Synonyms: leelamine
Molecular Weight (g/mol): 286.48
Molecular Formula: C20H32N
InChi Key: JVVXZOOGOGPDRZ-SLFFLAALSA-O
SMILES: CC(C)C1=CC=C2C(CC[C@H]3[C@](C)(C[NH3+])CCC[C@]23C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)