accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNNHH2NONOOHOOHOOHOOHO(E)(E)

CAS RN 144675-97-8

IUPAC Name: bis((2E)-but-2-enedioic acid); 6,9-bis[(2-aminoethyl)amino]-5H,10H-benzo[g]isoquinoline-5,10-dione
Synonyms: bis(fumaric acid); pixantrone
Molecular Weight (g/mol): 557.52
Molecular Formula: C25H27N5O10
InChi Key: SVAGFBGXEWPNJC-LVEZLNDCSA-N
SMILES: OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.NCCNC1=CC=C(NCCN)C2=C1C(=O)C1=CC=NC=C1C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results