accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHOONa

CAS RN 144693-65-2

IUPAC Name: sodium 4-ethynylbenzoate
Synonyms: sodium 4-ethynylbenzoate
Molecular Weight (g/mol): 168.13
Molecular Formula: C9H5NaO2
InChi Key: VXBLAFDQGACQAR-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC=C(C=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)