accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14483-17-1

H2OH2OH2OH2OH2OPrONOOONOOONOO

CAS RN 14483-17-1

IUPAC Name: praseodymium(3+) pentahydrate trinitrate
Synonyms: praseodymium(3+) pentahydrate trinitrate
Molecular Weight (g/mol): 417
Molecular Formula: H10N3O14Pr
InChi Key: PZVBXGDSTWEFMC-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results