CAS RN 14484-47-0
IUPAC Name:
2-[(1S,2S,4R,8S,9S,11S,12S,13R)-11-hydroxy-6,9,13-trimethyl-16-oxo-5-oxa-7-azapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-6,14,17-trien-8-yl]-2-oxoethyl acetate
Synonyms:
deflazacort galenpharma brand of deflazacort deflazacort (substance) lantadin cahill may roberts brand of deflazacort deflazacort (product) dezacor aventis brand of deflazacort azacort hoechst brand of deflazacort
Molecular Weight (g/mol):
441.52
Molecular Formula:
C25H31NO6
InChi Key:
FBHSPRKOSMHSIF-GRMWVWQJSA-N
SMILES:
CC(=O)OCC(=O)[C@@]12N=C(C)O[C@@H]1C[C@H]1[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]21C
1
–
1
of
1
results
1
Deflazacort 98.0+%, TCI America™
CAS: 14484-47-0 Molecular Formula: C25H31NO6 Molecular Weight (g/mol): 441.524 MDL Number: MFCD00866106 InChI Key: FBHSPRKOSMHSIF-GRMWVWQJSA-N PubChem CID: 189821 SMILES: CC1=NC2(C(O1)CC3C2(CC(C4C3CCC5=CC(=O)C=CC45C)O)C)C(=O)COC(=O)C