accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFFF

CAS RN 144851-61-6

IUPAC Name: 2-fluoro-6-(trifluoromethyl)aniline
Synonyms: 2-fluoro-6-(trifluoromethyl)aniline
Molecular Weight (g/mol): 179.12
Molecular Formula: C7H5F4N
InChi Key: CQSFHEFEKDRLKE-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results