CAS RN 1449131-72-9
IUPAC Name:
{4-fluoro-3-[(pyridin-2-yl)carbamoyl]phenyl}boronic acid
Synonyms:
4-fluoro-3-[(pyridin-2-yl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol):
260.03
Molecular Formula:
C12H10BFN2O3
InChi Key:
DJNCGHBHBJUZKA-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(C(=O)NC2=CC=CC=N2)=C(F)C=C1
1
–
1
of
1
results
1
4-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™
CAS: 1449131-72-9 Molecular Formula: C12H10BFN2O3 Molecular Weight (g/mol): 260.031 MDL Number: MFCD20040156 InChI Key: DJNCGHBHBJUZKA-UHFFFAOYSA-N Synonym: 4-fluoro-3-2-pyridylcarbamoyl benzeneboronic acid,4-fluoro-3-pyridin-2-yl carbamoyl phenylboronic acid PubChem CID: 73995690 IUPAC Name: [4-fluoro-3-(pyridin-2-ylcarbamoyl)phenyl]boronic acid SMILES: B(C1=CC(=C(C=C1)F)C(=O)NC2=CC=CC=N2)(O)O