accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1449132-35-7

IUPAC Name: (2-fluoro-5-{[(3-fluoro-4-methylphenyl)methyl]carbamoyl}phenyl)boronic acid
Synonyms: 2-fluoro-5-{[(3-fluoro-4-methylphenyl)methyl]carbamoyl}phenylboronic acid
Molecular Weight (g/mol): 305.09
Molecular Formula: C15H14BF2NO3
InChi Key: LZBLIQJRKZLMQM-UHFFFAOYSA-N
SMILES: CC1=C(F)C=C(CNC(=O)C2=CC(B(O)O)=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results