accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHNFHOOH

CAS RN 1449132-70-0

IUPAC Name: {5-[(cyclohexylmethyl)carbamoyl]-2-fluorophenyl}boronic acid
Synonyms: 5-(cyclohexylmethylcarbamoyl)-2-fluorophenylboronic acid
Molecular Weight (g/mol): 279.12
Molecular Formula: C14H19BFNO3
InChi Key: VRYJPOGKZNVFFH-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC(=C1)C(=O)NCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results