accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1449133-61-2

IUPAC Name: {4-[methyl(2-nitrophenyl)sulfamoyl]phenyl}boronic acid
Synonyms: 4-[methyl(2-nitrophenyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 336.13
Molecular Formula: C13H13BN2O6S
InChi Key: LXCUTVNBMYMJPZ-UHFFFAOYSA-N
SMILES: CN(C1=CC=CC=C1[N+]([O-])=O)S(=O)(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results