accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHNCH3CH3FHOOH

CAS RN 1449142-45-3

IUPAC Name: {5-[(2,6-dimethylphenyl)carbamoyl]-2-fluorophenyl}boronic acid
Synonyms: 5-[(2,6-dimethylphenyl)carbamoyl]-2-fluorophenylboronic acid
Molecular Weight (g/mol): 287.1
Molecular Formula: C15H15BFNO3
InChi Key: CHJDUXDVLGUAJJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC(=O)C1=CC(B(O)O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results