accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1449144-38-0

IUPAC Name: {2-fluoro-5-[(2-methoxy-4-nitrophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-fluoro-5-[(2-methoxy-4-nitrophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 334.07
Molecular Formula: C14H12BFN2O6
InChi Key: BYDYGPUXHRRBSV-UHFFFAOYSA-N
SMILES: COC1=C(NC(=O)C2=CC(B(O)O)=C(F)C=C2)C=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results