accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHNNFHOOH

CAS RN 1449144-50-6

IUPAC Name: {2-fluoro-5-[(pyridin-4-yl)carbamoyl]phenyl}boronic acid
Synonyms: 2-fluoro-5-[(pyridin-4-yl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 260.03
Molecular Formula: C12H10BFN2O3
InChi Key: QZOJQJVFSHJCBX-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results