accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1449144-56-2

IUPAC Name: {3-[(3-methylphenyl)sulfamoyl]phenyl}boronic acid
Synonyms: 3-[(3-methylphenyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 291.13
Molecular Formula: C13H14BNO4S
InChi Key: JATJSZDSJXABIZ-UHFFFAOYSA-N
SMILES: CC1=CC(NS(=O)(=O)C2=CC=CC(=C2)B(O)O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results