accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1449145-16-7

IUPAC Name: {3-[(2-methylpiperidin-1-yl)sulfonyl]phenyl}boronic acid
Synonyms: 3-(2-methylpiperidin-1-ylsulfonyl)phenylboronic acid
Molecular Weight (g/mol): 283.15
Molecular Formula: C12H18BNO4S
InChi Key: IZHIWXZQLZLQGQ-UHFFFAOYNA-N
SMILES: CC1CCCCN1S(=O)(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results