accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClBr

CAS RN 14495-51-3

IUPAC Name: 1-bromo-4-chloro-2-methylbenzene
Synonyms: 1-bromo-4-chloro-2-methylbenzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: RTIPTGMVQIIMKL-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)