accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 144981-85-1

IUPAC Name: 2-iodo-9,9-dimethyl-9H-fluorene
Synonyms: 2-iodo-9,9-dimethylfluorene
Molecular Weight (g/mol): 320.17
Molecular Formula: C15H13I
InChi Key: DVLSJPCXPNKPRJ-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(I)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results