accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOHHOOCH2OH

CAS RN 144985-19-3

IUPAC Name: 2-(hydroxymethyl)-6-(4-methoxyphenoxy)-4-(prop-2-en-1-yloxy)oxane-3,5-diol
Synonyms: 2-(hydroxymethyl)-6-(4-methoxyphenoxy)-4-(prop-2-en-1-yloxy)oxane-3,5-diol
Molecular Weight (g/mol): 326.35
Molecular Formula: C16H22O7
InChi Key: DXDVGRALJDFCOJ-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(CO)C(O)C(OCC=C)C2O)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)