accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3CSiCH3CH3H3C

CAS RN 1450-14-2

IUPAC Name: hexamethyldisilane
Synonyms: hexamethyldisilane
Molecular Weight (g/mol): 146.38
Molecular Formula: C6H18Si2
InChi Key: NEXSMEBSBIABKL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)[Si](C)(C)C
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (6)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (5)