accessibility menu, dialog, popup

UserName

Viewing profile as user:
SiSi

CAS RN 1450-23-3

IUPAC Name: hexaphenyldisilane
Synonyms: hexaphenyldisilane
Molecular Weight (g/mol): 518.81
Molecular Formula: C36H30Si2
InChi Key: ZMHATUZXFSOVSC-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results