accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHH3CO

CAS RN 1450-72-2

IUPAC Name: 1-(2-hydroxy-5-methylphenyl)ethan-1-one
Synonyms: 2-acetyl-4-methylphenol
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: YNPDFBFVMJNGKZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C)=CC=C1O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)