accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBrHO

CAS RN 1450-75-5

IUPAC Name: 1-(5-bromo-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(5-bromo-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: HQCCNFFIOWYINW-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Br)=CC=C1O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)