accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHNOHSOHOONH2NHN

CAS RN 1450-93-7

IUPAC Name: bis(1H-imidazol-2-amine); sulfuric acid
Synonyms: bis(1H-imidazol-2-amine); sulfuric acid
Molecular Weight (g/mol): 264.26
Molecular Formula: C6H12N6O4S
InChi Key: KUWRLKJYNASPQZ-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=NC=CN1.NC1=NC=CN1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)