accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14508-49-7

IUPAC Name: 2-chloropyrazine
Synonyms: 2-chloropyrazine
Molecular Weight (g/mol): 114.53
Molecular Formula: C4H3ClN2
InChi Key: GELVZYOEQVJIRR-UHFFFAOYSA-N
SMILES: ClC1=CN=CC=N1
Percent Purity 
  • (2)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)