accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1450841-27-6

IUPAC Name: 2,6-bis({1,1-diphenyl-hexahydropyrrolo[1,2-c][1,3]oxazol-3-yl})pyridine
Synonyms: 2,6-bis({1,1-diphenyl-tetrahydro-3H-pyrrolo[1,2-c][1,3]oxazol-3-yl})pyridine
Molecular Weight (g/mol): 605.78
Molecular Formula: C41H39N3O2
InChi Key: UFOJHPHCTXHMFQ-UHFFFAOYNA-N
SMILES: C1CC2N(C1)C(OC2(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC(=N1)C1OC(C2CCCN12)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results