accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14510-06-6

IUPAC Name: 8-hydroxyquinoline-2-carbaldehyde
Synonyms: 8-hydroxyquinoline-2-carbaldehyde
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: SLBPIHCMXPQAIQ-UHFFFAOYSA-N
SMILES: OC1=C2N=C(C=O)C=CC2=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)