accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOONNSOOFFF

CAS RN 145100-50-1

IUPAC Name: 1,1,1-trifluoro-N-(pyridin-2-yl)-N-trifluoromethanesulfonylmethanesulfonamide
Synonyms: 1,1,1-trifluoro-N-(pyridin-2-yl)-N-trifluoromethanesulfonylmethanesulfonamide
Molecular Weight (g/mol): 358.23
Molecular Formula: C7H4F6N2O4S2
InChi Key: DXLQEJHUQKKSRB-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)N(C1=CC=CC=N1)S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)