CAS RN 1451046-72-2
CAS RN 1451046-72-2
IUPAC Name:
(2S)-6-[(2E)-but-2-enamido]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms:
(2S)-6-[(2E)-but-2-enamido]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol):
436.51
Molecular Formula:
C25H28N2O5
InChi Key:
WFNXIABNUWPOIE-INZNARQDSA-N
SMILES:
C\C=C\C(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Lys Crotonyl -OH, Advanced ChemTech
CAS: 1451046-72-2 Molecular Formula: C25H28N2O5 Molecular Weight (g/mol): 436.51 InChI Key: WFNXIABNUWPOIE-INZNARQDSA-N IUPAC Name: (2S)-6-[(2E)-but-2-enamido]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid SMILES: C\C=C\C(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O