accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 145129-54-0

IUPAC Name: methyl 2,5-dichlorothiophene-3-carboxylate
Synonyms: methyl 2,5-dichlorothiophene-3-carboxylate
Molecular Weight (g/mol): 211.06
Molecular Formula: C6H4Cl2O2S
InChi Key: VCXPHMKCDRPIDG-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)SC(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)