accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14516-46-2

IUPAC Name: dipotassium palladiumtetracarbonitrile
Synonyms: dipotassium palladiumtetracarbonitrile
Molecular Weight (g/mol): 288.69
Molecular Formula: C4K2N4Pd
InChi Key: NKYOTIKKMTWAOI-UHFFFAOYSA-N
SMILES: [K+].[K+].N#C[Pd](C#N)(C#N)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results