CAS RN 1452-16-0
IUPAC Name:
1,3-thiazole-2-carbonitrile
Synonyms:
1,3-thiazole-2-carbonitrile
Molecular Weight (g/mol):
110.13
Molecular Formula:
C4H2N2S
InChi Key:
ZIRGWUZHKJDYKT-UHFFFAOYSA-N
SMILES:
N#CC1=NC=CS1
1
–
1
of
1
results
1
Promotions Available
2-Cyanothiazole, 95%
2-Cyanothiazole, 95%
CAS: 1452-16-0 Molecular Formula: C4H2N2S Molecular Weight (g/mol): 110.134 MDL Number: MFCD09702472 InChI Key: ZIRGWUZHKJDYKT-UHFFFAOYSA-N Synonym: 2-cyanothiazole,thiazole-2-carbonitrile,2-thiazolecarbonitrile,2-cyanothiazol PubChem CID: 15111480 IUPAC Name: 1,3-thiazole-2-carbonitrile SMILES: C1=CSC(=N1)C#N