accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNON

CAS RN 1452-63-7

IUPAC Name: pyridine-2-carbohydrazide
Synonyms: pyridine-2-carbohydrazide
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: BAQLNPIEFOYKNB-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results