accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NON

CAS RN 1452-77-3

IUPAC Name: pyridine-2-carboxamide
Synonyms: pyridine-2-carboxamide picolinoylamide α-picolinamide
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: IBBMAWULFFBRKK-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (5)
Boiling Point 
  • (1)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)