accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CH3COSNH3CH3COSNH3CCH3CH3CH3H3COSNCH3CH3CH3CH3CH3CH3CH3H3COSNCH3H3C

CAS RN 145237-46-3

IUPAC Name: N,N-dimethyl-2-({5,11,17,23-tetra-tert-butyl-26,27,28-tris[(dimethylcarbamothioyl)methoxy]pentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl}oxy)ethanethioamide
Synonyms: N,N-dimethyl-2-({5,11,17,23-tetra-tert-butyl-26,27,28-tris[(dimethylcarbamothioyl)methoxy]pentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl}oxy)ethanethioamide
Molecular Weight (g/mol): 1053.6
Molecular Formula: C60H84N4O4S4
InChi Key: CDVPPFVKRDRQTO-UHFFFAOYSA-N
SMILES: CN(C)C(=S)COC1=C2CC3=CC(=CC(CC4=C(OCC(=S)N(C)C)C(CC5=C(OCC(=S)N(C)C)C(CC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(=S)N(C)C)C(C)(C)C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results