accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CHOOH

CAS RN 145240-28-4

IUPAC Name: (4-butylphenyl)boronic acid
Synonyms: 4-butylphenylboronic acid
Molecular Weight (g/mol): 178.04
Molecular Formula: C10H15BO2
InChi Key: UGZUUTHZEATQAM-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results