CAS RN 145243-96-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Butalbital-D5 Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
A certified analytical reference solution and internal standard applicable for use in butalbital testing methods by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing or forensic analysis.