accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14526-22-8

IUPAC Name: iron(3+) tris(1,1,1-trifluoro-2,4-dioxopentan-3-ide)
Synonyms: iron(3+) tris(1,1,1-trifluoro-2,4-dioxopentan-3-ide)
Molecular Weight (g/mol): 515.09
Molecular Formula: C15H12F9FeO6
InChi Key: PDLKINIVWBCDRU-UHFFFAOYSA-N
SMILES: [Fe+3].CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F.CC(=O)[CH-]C(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results