accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHN

CAS RN 1453-58-3

IUPAC Name: 5-methyl-1H-pyrazole
Synonyms: 3-methyl-2H-pyrazole
Molecular Weight (g/mol): 82.11
Molecular Formula: C4H6N2
InChi Key: XKVUYEYANWFIJX-UHFFFAOYSA-N
SMILES: CC1=CC=NN1
Boiling Point 
  • (2)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (6)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)