accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1453071-47-0

IUPAC Name: 3-(9-cyclopropyl-N-hydroxynonanamido)propanoic acid
Synonyms: 3-(9-cyclopropyl-N-hydroxynonanamido)propanoic acid
Molecular Weight (g/mol): 285.38
Molecular Formula: C15H27NO4
InChi Key: NEHSERYKENINRH-UHFFFAOYSA-N
SMILES: ON(CCC(O)=O)C(=O)CCCCCCCCC1CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results