CAS RN 145343-76-6
IUPAC Name:
2-chloro-4-iodobenzoic acid
Synonyms:
2-chloro-4-iodobenzoic acid
Molecular Weight (g/mol):
282.46
Molecular Formula:
C7H4ClIO2
InChi Key:
KVFAMLOGLYILKM-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(Cl)C=C(I)C=C1
1
–
1
of
1
results
1
2-Chloro-4-iodobenzoic Acid 98.0+%, TCI America™
CAS: 145343-76-6 Molecular Formula: C7H4ClIO2 Molecular Weight (g/mol): 282.461 MDL Number: MFCD05863523 InChI Key: KVFAMLOGLYILKM-UHFFFAOYSA-N PubChem CID: 821264 IUPAC Name: 2-chloro-4-iodobenzoic acid SMILES: C1=CC(=C(C=C1I)Cl)C(=O)O