accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1454-84-8

H3COH

CAS RN 1454-84-8

IUPAC Name: nonadecan-1-ol
Synonyms: nonadecanol
Molecular Weight (g/mol): 284.53
Molecular Formula: C19H40O
InChi Key: XGFDHKJUZCCPKQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results