accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNCH3H3CONHNONNHNNHO

CAS RN 145455-35-2

IUPAC Name: 3-[(2-hydroxypropyl)amino]-3-methyl-N-(2-oxo-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)butanamide
Synonyms: 3-[(2-hydroxypropyl)amino]-3-methyl-N-(2-oxo-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-4,5-dihydro-3H-1-benzazepin-3-yl)butanamide
Molecular Weight (g/mol): 567.69
Molecular Formula: C32H37N7O3
InChi Key: AYBCFPXLXOLPIZ-UHFFFAOYNA-N
SMILES: CC(O)CNC(C)(C)CC(=O)NC1CCC2=CC=CC=C2N(CC2=CC=C(C=C2)C2=CC=CC=C2C2=NNN=N2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results