accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14548-31-3

IUPAC Name: hex-1-en-5-yne
Synonyms: hex-1-en-5-yne
Molecular Weight (g/mol): 80.13
Molecular Formula: C6H8
InChi Key: GHGNRQPVGKFJIR-UHFFFAOYSA-N
SMILES: C=CCCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results